Potassium tricyanomethanide
Supplier: Thermo Scientific Chemicals
|
Danger
|
20702-03
20702-06
CAAA20702-03EA
339.7
CAD
CAAA20702-03
CAAA20702-06
Potassium tricyanomethanide
Potassium tricyanomethanide
CAS No.: 34171-69-2
Molecular Formula: C4KN3
Formula Weight: 129.16
Physical Form: Crystalline
Appearance: Off-white
UN No.: 3439
MDL No.: MFCD00058850
Formula:
C₄KN₃ Storage Temperature: Ambient |
MDL Number:
MFCD00058850 CAS Number: 34171-69-2 UN: 3439 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...