Iron(III) hydroxide, α-phase
Supplier: Thermo Scientific Chemicals
Applications: In purifying water, as absorbent in chemical processes, as a pigment, and as a catalyst
Insoluble in water and alcohol. Soluble in mineral acids
CAS No.: 20344-49-4
Molecular Formula: FeO(OH)
Formula Weight: 88.85
Physical Form: Powder
Density: 3.4-3.9
Odor: Odorless
Solubility: Insoluble in water and alcohol. Soluble in mineral acids
MDL No.: MFCD00064782
Applications: In purifying water, as absorbent in chemical processes, as a pigment, and as a catalyst
Formula:
FeO(OH) Density: 3.4…3.9 Storage Temperature: Ambient |
MDL Number:
MFCD00064782 CAS Number: 20344-49-4 EINECS: 243-746-4 Merck Index: 14,4025 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...