1-Cyclohexene-1-acetic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Cyclohexenylacetic acid
19462-03
19462-06
19462-14
CAAA19462-03EA
59.25
CAD
CAAA19462-03
CAAA19462-06
CAAA19462-14
1-Cyclohexene-1-acetic acid
1-Cyclohexene-1-acetic acid
CAS No.: 18294-87-6
Molecular Formula: C8H12O2
Formula Weight: 140.18
Physical Form: Liquid/low melting solid
Density: 1.04
Refractive Index: 1.4775
Melting Point: 33°
Boiling Point: 140-142°/25mm
MDL No.: MFCD00015482
Formula:
C₈H₁₂O₂ MW: 140.182 g/mol Boiling Pt: 140…142 °C (25 mmHg) Melting Pt: 33 °C Density: 1,04 Storage Temperature: Ambient |
MDL Number:
MFCD00015482 CAS Number: 18294-87-6 EINECS: 242-172-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...