Nickel chromite

Supplier: Thermo Scientific Chemicals
Danger

19424-14 19424-22
CAAA19424-14EA 148.99 CAD
CAAA19424-14 CAAA19424-22
Nickel chromite
Nickel chromite

Contains approximately 10% NiCrO₃. Dehydrogenation catalyst


CAS No.: 12018-18-7 Molecular Formula: Cr2NiO4 Formula Weight: 226.70 Physical Form: -100 Mesh Powder Odor: Odorless MDL No.: MFCD00064806

Formula: Cr₂NiO₄
Storage Temperature: Ambient
MDL Number: MFCD00064806
CAS Number: 12018-18-7
EINECS: 234-636-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR