Copper(I) bromide dimethyl sulfide complex
Supplier: Thermo Scientific Chemicals
|
Warning
|
19368-04
19368-09
19368-18
CAAA19368-04EA
42.34
CAD
CAAA19368-04
CAAA19368-09
CAAA19368-18
Copper(I) bromide dimethyl sulfide complex
Copper (I) bromide dimethyl sulphide complex
CAS No.: 54678-23-8
Molecular Formula: CuBr.S(CH3)2
Formula Weight: 205.58
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: Pale brown or pale green
Melting Point: 124-129° dec.
UN No.: 3335
MDL No.: MFCD00043295
Formula:
C₂H₆BrCuS MW: 205.58 Melting Pt: 124…129 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00043295 CAS Number: 54678-23-8 EINECS: 259-292-5 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...