Tetrabutylammonium acetate
Supplier: Thermo Scientific Chemicals
|
Warning
|
19279-03
19279-06
CAAA19279-03EA
49.61
CAD
CAAA19279-03
CAAA19279-06
Tetrabutylammonium acetate
Tetrabutylammonium acetate
CAS No.: 10534-59-5
Molecular Formula: C18H39NO2
Formula Weight: 301.51
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 114-115°
MDL No.: MFCD00043208
Formula:
C₁₈H₃₉NO₂ Melting Pt: 114…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043208 CAS Number: 10534-59-5 EINECS: 234-101-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...