Bis(triphenylphosphine)nickel(II) iodide
Supplier: Thermo Scientific Chemicals
|
Danger
|
18869-04
18869-09
CAAA18869-04EA
87.85
CAD
CAAA18869-04
CAAA18869-09
Bis(triphenylphosphine)nickel(II) iodide
Bis(triphenylphosphine)nickel (II) iodide
CAS No.: 82308-17-6
Molecular Formula: C36H30I2NiP2
Formula Weight: 837.08
Physical Form: Crystalline powder
Sensitivity: Air and Light Sensitive
Appearance: Dark brown
MDL No.: MFCD00015866
Formula:
C₃₆H₃₀I₂NiP₂ Storage Temperature: Ambient |
MDL Number:
MFCD00015866 CAS Number: 82308-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...