Copper(II) methoxide ≥98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
18861-03
18861-06
18861-14
CAAA18861-03EA
52.61
CAD
CAAA18861-03
CAAA18861-06
CAAA18861-14
Copper(II) methoxide ≥98% (metals basis)
Copper(II) methoxide
Purity: 98%
CAS No.: 1184-54-9
Molecular Formula: C2H6CuO2
Formula Weight: 125.61
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Green
Odor: Alcohol-like
Melting Point: ca 206° dec.
UN No.: 3181
MDL No.: MFCD00015611
Formula:
Cu(CH₃O)₂ Melting Pt: ∼ 206 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00015611 CAS Number: 1184-54-9 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...