Iron(III) meso-tetraphenylporphine chloride

Supplier: Thermo Scientific Chemicals

18829-03 18829-MD
CAAA18829-03EA 290.72 CAD
CAAA18829-03 CAAA18829-MD
Iron(III) meso-tetraphenylporphine chloride
Iron(III) meso-tetraphenylporphine chloride

CAS No.: 16456-81-8 Molecular Formula: C44H28ClFeN4 Formula Weight: 704.04 Physical Form: Crystalline MDL No.: MFCD00012152

Formula: C₄₄H₃₀Cl₃FeN₄
Storage Temperature: Ambient
MDL Number: MFCD00012152
CAS Number: 16456-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR