Zinc trifluoroacetate hydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
18686-14
18686-22
CAAA18686-14EA
191.18
CAD
CAAA18686-14
CAAA18686-22
Zinc trifluoroacetate hydrate
Zinc trifluoroacetate hydrate
CAS No.: 207801-31-8
Molecular Formula: Zn(OOCCF3)2.xH2O
Formula Weight: 291.40(anhy)
Physical Form: Powder/chunks
Sensitivity: Hygroscopic
UN No.: 3077
MDL No.: MFCD00061595
Formula:
C₄H₂F₆O₅Zn Storage Temperature: Ambient |
MDL Number:
MFCD00061595 CAS Number: 207801-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...