Molybdenum(II) acetate dimer

Supplier: Thermo Scientific Chemicals

18239-03 18239-06
CAAA18239-03EA 122.77 CAD
CAAA18239-03 CAAA18239-06
Molybdenum(II) acetate dimer
Molybdenum(II) acetate dimer

CAS No.: 14221-06-8 Molecular Formula: [Mo(OCOCH3)2]2 Formula Weight: 428.06 Physical Form: Crystalline Sensitivity: Air Sensitive MDL No.: MFCD00013395

Formula: C₈H₁₂Mo₂O₈
MW: 428.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013395
CAS Number: 14221-06-8
EINECS: 238-089-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR