Magnesium phosphate pentahydrate (33% P₂O₅)
Supplier: Thermo Scientific Chemicals
|
Warning
|
18219-A1
18219-A7
CAAA18219-A1EA
120.4
CAD
CAAA18219-A1
CAAA18219-A7
Magnesium phosphate pentahydrate (33% P₂O₅)
Magnesium phosphate pentahydrate
CAS No.: 10233-87-1
Molecular Formula: Mg3(PO4)2.5H2O
Formula Weight: 352.93 (262.86anhy)
Physical Form: Crystalline
Appearance: White
MDL No.: MFCD00150252
Formula:
Mg₃(PO₄)₂.5H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150252 CAS Number: 10233-87-1 EINECS: 231-824-0 Merck Index: 14,5683 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...