Lithium chloride/potassium chloride eutectic
Supplier: Thermo Scientific Chemicals
|
Warning
|
18206-18
18206-30
CAAA18206-18EA
202.24
CAD
CAAA18206-18
CAAA18206-30
Lithium chloride/potassium chloride eutectic
Lithium chloride/potassium chloride eutectic
CAS No.: 65567-96-6
Molecular Formula: LiCl.KCl
Physical Form: -10 Mesh
Density: 2.02
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 357°
MDL No.: MFCD00144623
Formula:
Cl₂KLi MW: 116.938 g/mol Melting Pt: 357 °C Density: 2,02 Storage Temperature: Ambient |
MDL Number:
MFCD00144623 CAS Number: 65567-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...