Gallium tin alloy ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
18161-03
18161-09
18161-18
CAAA18161-03EA
39.66
CAD
CAAA18161-03
CAAA18161-09
CAAA18161-18
Gallium tin alloy ≥99.99% (metals basis)
Gallium tin alloy
Purity: 99.99%
Molecular Formula: Ga:Sn; 92:8 wt%
Physical Form: Liquid
Appearance: Silver
Odor: Odorless
Melting Point: ≈25°
UN No.: 2803
MDL No.: MFCD00144390
Formula:
Ga:Sn; 92:8 % Melting Pt: ∼25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00144390 UN: 2803 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...