Zinc citrate dihydrate (≥31% Zn), Reagent Grade
Supplier: Thermo Scientific Chemicals
18129-30
18129-A1
CAAA18129-30EA
79.95
CAD
CAAA18129-30
CAAA18129-A1
Zinc citrate dihydrate (≥31% Zn), Reagent Grade
Zinc citrate dihydrate
Soluble in dilute mineral acids and alkali hydroxides. Slightly soluble in water
CAS No.: 5990-32-9
Molecular Formula: C12H10O14Zn3.2H2O
Formula Weight: 610.35 (574.32anhy)
Physical Form: Powder
Odor: Odorless
Melting Point: 334° dec.
Solubility: Soluble in dilute mineral acids and alkali hydroxides. Slightly soluble in water
MDL No.: MFCD00150587
Formula:
C₁₂H₁₄O₁₆Zn₃ Melting Pt: 334 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150587 CAS Number: 5990-32-9 EINECS: 208-901-2 Merck Index: 14,10134 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...