Cobalt aluminate (39 - 41% Co)

Supplier: Thermo Scientific Chemicals
Warning

17686-22 17686-36
CAAA17686-22EA 71.26 CAD
CAAA17686-22 CAAA17686-36
Cobalt aluminate (39 - 41% Co)
Cobalt aluminate

CAS No.: 12672-27-4 Molecular Formula: CoAl2O4 (approximate) Formula Weight: 176.89 Physical Form: -200 Mesh Powder Odor: Odorless MDL No.: MFCD00016016

Formula: Al₂Co₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00016016
CAS Number: 12672-27-4
EINECS: 235-762-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR