Triallyl phosphite
Supplier: Thermo Scientific Chemicals
|
Danger
|
17484-09
17484-18
17484-30
CAAA17484-09EA
126.72
CAD
CAAA17484-09
CAAA17484-18
CAAA17484-30
Triallyl phosphite
Triallyl phosphite
CAS No.: 102-84-1
Molecular Formula: C9H15O3P
Formula Weight: 202.19
Flash Point: 86°(187°F)
Physical Form: Liquid
Sensitivity: Air and Moisture Sensitive
Appearance: Colorless
Odor: Strong
Boiling Point: 84°/12mm
Beilstein Reference: 1705535
UN No.: 3278
MDL No.: MFCD00014948
Formula:
C₉H₁₅O₃P Boiling Pt: 84 °C (12 mmHg) Flash Pt: 86 °C (187 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014948 CAS Number: 102-84-1 EINECS: 203-060-8 UN: 3278 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...