Lithium tetrachloropalladate(II)

Supplier: Thermo Scientific Chemicals
Warning

17316-03 17316-06
CAAA17316-03EA 251.22 CAD
CAAA17316-03 CAAA17316-06
Lithium tetrachloropalladate(II)
Lithium tetrachloropalladate(II)

CAS No.: 15525-45-8 Molecular Formula: Li2PdCl4 Formula Weight: 262.09 Physical Form: Crystalline Sensitivity: Moisture Sensitive MDL No.: MFCD00011082

Formula: Cl₄Li₂Pd
MW: 262.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011082
CAS Number: 15525-45-8
EINECS: 239-567-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR