Tris(trimethylsilyl)phosphite ≥96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(trimethylsilyl)phosphite

17159-02 17159-04
CAAA17159-02EA 36.81 CAD
CAAA17159-02 CAAA17159-04
Tris(trimethylsilyl)phosphite ≥96%
Tris(trimethylsilyl) phosphite

Purity: 96% CAS No.: 1795-31-9 Molecular Formula: [(CH3)3SiO]3P Formula Weight: 298.54 Physical Form: Liquid Density: 0.893 Refractive Index: 1.409 Sensitivity: Air and Moisture Sensitive Boiling Point: 80°/8mm MDL No.: MFCD00015588

Formula: C₉H₂₇O₃PSi₃
MW: 298.54 g/mol
Boiling Pt: 80 °C (8 mmHg)
Density: 0.893
Storage Temperature: Ambient
MDL Number: MFCD00015588
CAS Number: 1795-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR