Bis(cyclopentadienyl)cobalt

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cobaltocene, Bis(cyclopentadienyl)cobalt(II)

16857-03 16857-06 16857-14
CAAA16857-03EA 95.27 CAD
CAAA16857-03 CAAA16857-06 CAAA16857-14
Bis(cyclopentadienyl)cobalt
Bis(cyclopentadienyl)cobalt

CAS No.: 1277-43-6 Molecular Formula: C10H10Co Formula Weight: 189.12 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive Melting Point: 176-180° dec. UN No.: 3181 MDL No.: MFCD00013749

Formula: C₁₀H₁₀Co
MW: 189.12 g/mol
Melting Pt: 176…180 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00013749
CAS Number: 1277-43-6
EINECS: 215-061-0
UN: 3181
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR