Bis(4-bromophenyl)ether ≥99%
Supplier: Thermo Scientific Chemicals
Synonyms:
4,4'-Dibromodiphenyl Ether, 4,4'-Oxybis(bromobenzene), 4 4'-OXYBIS(BROMOBENZENE)
16684-06
16684-14
CAAA16684-06EA
69.68
CAD
CAAA16684-06
CAAA16684-14
Bis(4-bromophenyl)ether ≥99%
Bis(4-bromophenyl)ether
Purity: 99%
CAS No.: 2050-47-7
Molecular Formula: C12H8Br2O
Formula Weight: 328.01
Physical Form: Crystalline
Appearance: White
Melting Point: 56-60°
Boiling Point: 338-340°
MDL No.: MFCD00000095
Formula:
C₁₂H₈Br₂O MW: 328.00 g/mol Boiling Pt: 338…340 °C Melting Pt: 56…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000095 CAS Number: 2050-47-7 EINECS: 218-090-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...