Trimethylolpropane triacrylate stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TMPTA, 1,1,1-Tris(acryloyloxymethyl)propane

16445-22 16445-36 16445-A3
CAAA16445-22EA 61.78 CAD
CAAA16445-22 CAAA16445-36 CAAA16445-A3
Trimethylolpropane triacrylate stabilized
Trimethylolpropane triacrylate

CAS No.: 15625-89-5 Molecular Formula: (CH2=CHCO2CH2)3CCH2CH3 Formula Weight: 296.31 Flash Point: 87°(188°F) Physical Form: Liquid Density: 1.10 Refractive Index: 1.4735 Odor: Pungent Boiling Point: 200°/1mm MDL No.: MFCD00008628

Formula: C₁₅H₂₀O₆
MW: 296.32 g/mol
Boiling Pt: >200 °C (1 mmHg)
Density: 1,10
Flash Pt: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008628
CAS Number: 15625-89-5
EINECS: 239-701-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR