Tetrabutylammonium hydroxide 55% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

16198-18 16198-30
CAAA16198-18EA 106.18 CAD
CAAA16198-18 CAAA16198-30
Tetrabutylammonium hydroxide 55% (w/w) in aqueous solution
Tetrabutylammonium hydroxide

Applications: reagent for ion-pair extractions


CAS No.: 2052-49-5 Molecular Formula: C16H37NO Formula Weight: 259.48 Flash Point: None Physical Form: Liquid Density: 0.97 Refractive Index: 1.435 Sensitivity: Air Sensitive Appearance: Colorless to light yellow pH: 14 Beilstein Reference: 3571228 UN No.: 3267 MDL No.: MFCD00009425


As a reagent for ion-pair extractions

Formula: (C₄H₉)₄NOH
Density: 0.97
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR