Copper(II) gluconate

Supplier: Thermo Scientific Chemicals

Synonyms: Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate

15325-18 15325-30 15325-A1
CAAA15325-18EA 51.19 CAD
CAAA15325-18 CAAA15325-30 CAAA15325-A1
Copper(II) gluconate
Copper(II) gluconate

CAS No.: 527-09-3 Molecular Formula: [HOCH2[CH(OH)]4CO2]2Cu Formula Weight: 453.85 Physical Form: Powder Melting Point: 155-157° MDL No.: MFCD00075297

Formula: C₆H₁₂O₇
MW: 453.84 g/mol
Melting Pt: 155…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00075297
CAS Number: 527-09-3
EINECS: 208-408-2
Merck Index: 14,2640

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR