Neodymium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

15301-18 15301-30
CAAA15301-18EA 252.8 CAD
CAAA15301-18 CAAA15301-30
Neodymium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis)
Neodymium(III) carbonate hydrate

Purity: 99.9% CAS No.: 38245-38-4 Molecular Formula: Nd2(CO3)3.xH2O Formula Weight: 468.51(anhy) Physical Form: Powder MDL No.: MFCD00150426

Formula: Nd₂(CO₃)₃.xH₂O
MW: 468.51 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150426
CAS Number: 38245-38-4
EINECS: 227-579-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR