Lanthanum triacetate sesquihydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Lanthanum(III) acetate sesquihydrate

15299-22 15299-36
CAAA15299-22EA 98.91 CAD
CAAA15299-22 CAAA15299-36
Lanthanum triacetate sesquihydrate ≥99.9% (REO, rare earth oxide basis)
Lanthanum triacetate sesquihydrate

Purity: 99.9% CAS No.: 25721-92-0 Molecular Formula: La(OOCCH3)3.1.5H2O Formula Weight: 343.05 (316.03anhy) Physical Form: Lump/Powder Sensitivity: Hygroscopic MDL No.: MFCD00013037

Formula: C₆H₉LaO₆·1.5H₂O
MW: 343.05 g/mol (316.03 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00013037
CAS Number: 25721-92-0
EINECS: 213-034-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR