Cerium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

15295-22 15295-36
CAAA15295-22EA 140.78 CAD
CAAA15295-22 CAAA15295-36
Cerium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Cerium (III) carbonate hydrate

Insoluble in water. Soluble indilute mineral acids.


Purity: 99.9% CAS No.: 54451-25-1 Molecular Formula: Ce2(CO3)3.xH2O Formula Weight: 460.27(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Solubility: Insoluble in water. Soluble indilute mineral acids. Merck Reference: 14,1996 MDL No.: MFCD00149635

Formula: C₃Ce₂O₉·H₂O
MW: 478.274 g/mol
Melting Pt: 848 °C
Storage Temperature: Ambient
MDL Number: MFCD00149635
CAS Number: 54451-25-1
EINECS: 208-655-6
Merck Index: 14,1996

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR