Europium(III) oxalate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

15288-04 15288-09
CAAA15288-04EA 82.79 CAD
CAAA15288-04 CAAA15288-09
Europium(III) oxalate hydrate ≥99.9% (REO, rare earth oxide basis)
Europium(III) oxalate hydrate

Purity: 99.9% CAS No.: 152864-32-9 Molecular Formula: C6Eu2O12.xH2O Formula Weight: 567.98(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White UN No.: 3288 MDL No.: MFCD00150143

Formula: C₆Eu₂O₁₂·xH₂O
MW: 567.98 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150143
CAS Number: 152864-32-9
EINECS: 221-885-1
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR