Dysprosium(III) carbonate tetrahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
15286-09
15286-18
CAAA15286-09EA
76.47
CAD
CAAA15286-09
CAAA15286-18
CAAA15286-30
Dysprosium(III) carbonate tetrahydrate ≥99.9% (REO, rare earth oxide basis)
Dysprosium (III) carbonate tetrahydrate
Purity: 99.9%
CAS No.: 38245-35-1
Molecular Formula: Dy2(CO3)3.4H2O
Formula Weight: 577.06 (505.03anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00150229
Formula:
C₃Dy₂O₉·4H₂O MW: 577.088 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150229 CAS Number: 38245-35-1 EINECS: 225-770-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...