Dodecyl methyl sulfide

Supplier: Thermo Scientific Chemicals

14877-06 14877-14
CAAA14877-06EA 97.64 CAD
CAAA14877-06 CAAA14877-14
Dodecyl methyl sulfide
Dodecyl methyl sulphide

CAS No.: 3698-89-3 Molecular Formula: C13H28S Formula Weight: 216.41 Physical Form: Liquid Refractive Index: 1.4608 Odor: Sulphurous Boiling Point: 163-165°/19mm MDL No.: MFCD00015073

Formula: C₁₃H₂₈S
Boiling Pt: 163…165 °C (19 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00015073
CAS Number: 3698-89-3
EINECS: 223-028-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR