Yttrium(III) hexafluoro-2,4-pentanedionate
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Yttrium(III) hexafluoroacetylacetonate, Yttrium hexafluoroacetylacetonate
14717-03
14717-06
CAAA14717-03EA
76.63
CAD
CAAA14717-03
CAAA14717-06
Yttrium(III) hexafluoro-2,4-pentanedionate
Yttrium(III) hexafluoro-2,4-pentanedionate
CAS No.: 18911-76-7
Molecular Formula: C15H3F18O6Y
Formula Weight: 710.10
Physical Form: Crystalline
Melting Point: 166-170°
MDL No.: MFCD00064764
Formula:
C₁₅H₃F₁₈O₆Y MW: 710.10 g/mol Melting Pt: 166…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064764 CAS Number: 18911-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...