Silicon tetraacetate ≥97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14703-18
14703-30
CAAA14703-18EA
232.26
CAD
CAAA14703-18
CAAA14703-30
Silicon tetraacetate ≥97%
Silicon tetraacetate
CAS No.: 562-90-3
Molecular Formula: Si(OOCCH3)4
Formula Weight: 264.27
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 110°
Boiling Point: 160-170° dec.
UN No.: 3261
MDL No.: MFCD00026184
Formula:
C₈H₁₂O₈Si Boiling Pt: 160…170 °C (dec.) Melting Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026184 CAS Number: 562-90-3 EINECS: 209-239-7 UN: 3261 ADR: 8,II Merck Index: 14,8498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...