Manganese(II) oxalate dihydrate (≥30% Mn)

Supplier: Thermo Scientific Chemicals
Warning

14685-06 14685-18 14685-30 14685-A1
CAAA14685-06EA 29.55 CAD
CAAA14685-06 CAAA14685-18 CAAA14685-30 CAAA14685-A1
Manganese(II) oxalate dihydrate (≥30% Mn)
Manganese(II) oxalate dihydrate

Applications: paint and varnish drier
Soluble in dilute acids. Slightly soluble in water


CAS No.: 6556-16-7 Molecular Formula: MnC2O4.2H2O Formula Weight: 178.99 (142.96anhy) Physical Form: Powder/chunks Density: 2.453 Appearance: White Odor: Odorless Melting Point: 100° dec. Solubility: Soluble in dilute acids. Slightly soluble in water MDL No.: MFCD00150452


Applications: As a paint and varnish drier

Formula: C₂H₆MnO₆
MW: 181.002 g/mol
Melting Pt: 100 °C (dec.)
Density: 2.453
Storage Temperature: Ambient
MDL Number: MFCD00150452
CAS Number: 6556-16-7
EINECS: 211-367-3
Merck Index: 14,5734

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR