Lead(II) zirconate ≥99.7% (max. 1% Hf)

Supplier: Thermo Scientific Chemicals
Danger

14682-22 14682-36
CAAA14682-22EA 442.4 CAD
CAAA14682-22 CAAA14682-36
Lead(II) zirconate ≥99.7% (max. 1% Hf)
Lead(II) zirconate

Purity: 99.7% CAS No.: 12060-01-4 Molecular Formula: PbZrO3 Formula Weight: 346.42 Physical Form: -325 Mesh Powder Density: 7.0 Odor: Odorless UN No.: 3077 MDL No.: MFCD00016275

Formula: O₃PbZr
Density: 7.0
Storage Temperature: Ambient
MDL Number: MFCD00016275
CAS Number: 12060-01-4
EINECS: 235-039-4
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR