Calcium disilicide Ca ∼30% may contain up to 5% Fe, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
14676-22
14676-36
CAAA14676-22EA
89.74
CAD
CAAA14676-22
CAAA14676-36
Calcium disilicide Ca ∼30% may contain up to 5% Fe, tech.
Calcium disilicide
Insoluble in cold water, decomposes in hot water
CAS No.: 12013-56-8
Molecular Formula: CaSi2
Formula Weight: 96.25
Physical Form: Powder
Density: 2.5
Sensitivity: Moisture Sensitive
Appearance: Grey
Melting Point: ca 1000°
Solubility: Insoluble in cold water, decomposes in hot water
UN No.: 1405
MDL No.: MFCD00015977
Formula:
CaSi₂ Melting Pt: ∼ 1000 °C Density: 2.5 Storage Temperature: Ambient |
MDL Number:
MFCD00015977 CAS Number: 12013-56-8 EINECS: 234-588-7 UN: 1405 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...