Gallium indium tin alloy (62:22:16 w%), eutectic ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
14634-03
14634-09
14634-18
CAAA14634-03EA
64.15
CAD
CAAA14634-03
CAAA14634-09
CAAA14634-18
Gallium indium tin alloy (62:22:16 w%), eutectic ≥99.99% (metals basis)
Gallium Indium alloy (62:22:16 w%), eutectic
Purity: 99.99%
Molecular Formula: Ga:In:Sn; 62:22:16 wt%
Physical Form: Liquid
Density: 6.359
Appearance: Silvery white
Odor: Odorless
UN No.: 2803
MDL No.: MFCD00144388
Melting Pt:
10.7 °C Density: 6.359 Storage Temperature: Ambient |
MDL Number:
MFCD00144388 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...