Mineral oil, white (petroleum), 75 cSt

Supplier: Thermo Scientific Chemicals
Danger

14621-LV 14621-MQ
CAAA14621-2YEA 261.06 CAD
CAAA14621-2Y CAAA14621-LV CAAA14621-MQ
Mineral oil, white (petroleum), 75 cSt
White mineral oil (petroleum)

CAS No.: 8042-47-5 Flash Point: 110°(230°F) Physical Form: Oily liquid Appearance: Colorless Odor: Odorless MDL No.: MFCD00131611

Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00147539
CAS Number: 8042-47-5
EINECS: 232-455-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR