Lanthanum(III) isopropoxide La 40%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14616-02
14616-04
14616-09
CAAA14616-02EA
100.8
CAD
CAAA14616-02
CAAA14616-04
CAAA14616-09
Lanthanum(III) isopropoxide La 40%
Lanthanum (III) isopropoxide
CAS No.: 19446-52-7
Molecular Formula: C9H21LaO3
Formula Weight: 316.18
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: White
Odor: Alcohol-like
Melting Point: 120-128°
Boiling Point: 170°/0.04mm subl.
UN No.: 2925
MDL No.: MFCD00015305
Formula:
C₉H₂₁LaO₃ MW: 316.18 g/mol Boiling Pt: 170 °C (0.04 mmHg) (subl.) Melting Pt: 120…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015305 CAS Number: 19446-52-7 UN: 2925 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...