Praseodymium(III) acetate hydrate ≥99.9%
Supplier: Thermo Scientific Chemicals
Synonyms:
Praseodynium acetate hydrate
14611-14
14611-22
CAAA14611-14EA
57.83
CAD
CAAA14611-14
CAAA14611-22
Praseodymium(III) acetate hydrate ≥99.9%
Praseodymium(III) acetate hydrate
Purity: 99.9%
CAS No.: 6192-12-7
Molecular Formula: Pr(OOCCH3)3.xH2O
Formula Weight: 318.03(anhy)
Physical Form: Crystalline Aggregates
MDL No.: MFCD00150121
Formula:
Pr(OOCCH₃)₃.xH₂O MW: 318.03 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150121 CAS Number: 6192-12-7 EINECS: 228-242-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...