Samarium(III) acetate hydrate ≥99.9%

Supplier: Thermo Scientific Chemicals

14604-14 14604-22
CAAA14604-14EA 126.08 CAD
CAAA14604-14 CAAA14604-22
Samarium(III) acetate hydrate ≥99.9%
Samarium(III) acetate hydrate

Partial dehydration at 100°

Soluble in water


Purity: 99.9% CAS No.: 17829-86-6 Molecular Formula: Sm(OOCCH3)3.xH2O Formula Weight: 327.48(anhy) Physical Form: Crystalline Aggregates Density: 1.94 Sensitivity: Hygroscopic Odor: Odorless Boiling Point: 300° Solubility: Soluble in water MDL No.: MFCD00150124

Formula: Sm(OOCCH₃)₃·xH₂O
MW: 327.48 g/mol (anhy)
Boiling Pt: 300 °C
Density: 1.94
Storage Temperature: Ambient
MDL Number: MFCD00150124
CAS Number: 17829-86-6
EINECS: 233-950-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR