Samarium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
14601-14
14601-22
CAAA14601-14EA
74.89
CAD
CAAA14601-14
CAAA14601-22
Samarium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Samarium (III) sulphate octahydrate
Sparingly soluble in water
Purity: 99.9%
CAS No.: 13465-58-2
Molecular Formula: Sm2(SO4)3.8H2O
Formula Weight: 733.00 (588.88anhy)
Physical Form: Crystalline
Melting Point: 450° -8H2O
MDL No.: MFCD00011236
Formula:
O₁₂S₃Sm₂·8H₂O MW: 733.03 g/mol Melting Pt: 450 °C (-8H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00011236 CAS Number: 13465-58-2 EINECS: 237-211-4 Merck Index: 14,8347 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...