Holmium(III) acetate hydrate ≥99.9%, REacton®

Supplier: Thermo Scientific Chemicals
Warning

14591-09 14591-18
CAAA14591-04EA 66.96 CAD
CAAA14591-04 CAAA14591-09 CAAA14591-18
Holmium(III) acetate hydrate ≥99.9%, REacton®
Holmium(III) acetate hydrate

Purity: 99.9% CAS No.: 312619-49-1 Molecular Formula: Ho(OOCCH3)3.xH2O Formula Weight: 342.07(anhy) Physical Form: Crystalline Aggregates MDL No.: MFCD00150648

Formula: C₆H₉HoO₆·xH₂O
MW: 342.07 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150648
CAS Number: 312619-49-1
EINECS: 247-066-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR