Ytterbium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

14574-09 14574-18
CAAA14574-09EA 136.67 CAD
CAAA14574-09 CAAA14574-18
Ytterbium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium(III) carbonate hydrate

Purity: 99.9% Molecular Formula: Yb2(CO3)3.xH2O Formula Weight: 526.11(anhy) Physical Form: Crystalline Melting Point: dec. MDL No.: MFCD00050146

Formula: 3(CO₃)·Yb₂·xH₂O
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00798154
CAS Number: 5895-52-3
EINECS: 227-585-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR