Ytterbium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

14572-09 14572-18
CAAA14572-09EA 86.11 CAD
CAAA14572-09 CAAA14572-18
Ytterbium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium (III) sulphate octahydrate

Purity: 99.9% CAS No.: 10034-98-7 Molecular Formula: Yb2(SO4)3.8H2O Formula Weight: 778.38 (634.26anhy) Physical Form: Crystalline

Formula: O₁₂S₃Yb₂·8H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149887
CAS Number: 10034-98-7
EINECS: 236-727-7
Merck Index: 14,10105

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR