Yttrium(III) acetate tetrahydrate ≥99.9%

Supplier: Thermo Scientific Chemicals

14565-18 14565-30
CAAA14565-18EA 109.81 CAD
CAAA14565-18 CAAA14565-30
Yttrium(III) acetate tetrahydrate ≥99.9%
Yttrium(III) acetate tetrahydrate

Applications: In analytical chemistry
Soluble in hot water


Purity: 99.9% CAS No.: 85949-60-6 Molecular Formula: C6H9O6Y.4H2O Formula Weight: 338.10 (266.04anhy) Physical Form: Powder Density: 1.5 Appearance: White Melting Point: dec. Solubility: Soluble in hot water MDL No.: MFCD00013048


Applications: In analytical chemistry

Formula: C₆H₉O₆Y·4H₂O
Melting Pt: Dec.
Density: 1,5
Storage Temperature: Ambient
MDL Number: MFCD00013048
CAS Number: 85949-60-6
EINECS: 245-612-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR