Yttrium(III) sulfate octahydrate ≥97% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
14561-18
14561-30
CAAA14561-18EA
112.81
CAD
CAAA14561-18
CAAA14561-30
Yttrium(III) sulfate octahydrate ≥97% (REO, rare earth oxide basis)
Yttrium(III) sulfate octahydrate
Water solubility decreases with increasing temperature
Purity: 99.9%
CAS No.: 7446-33-5
Molecular Formula: Y2(SO4)3.8H2O
Formula Weight: 610.11 (465.99anhy)
Physical Form: Crystalline
Appearance: White to pale yellow
Odor: Odorless
Solubility: Water solubility decreases with increasing temperature
MDL No.: MFCD00149946
Formula:
H₁₆O₂₀S₃Y₂ Storage Temperature: Ambient |
MDL Number:
MFCD00149946 CAS Number: 7446-33-5 EINECS: 236-844-3 Merck Index: 14,10107 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...