Aluminum titanium oxide ≥99.5% (metals basis), powder -100 Mesh

Supplier: Thermo Scientific Chemicals
Warning

14484-14 14484-22
CAAA14484-14EA 125.61 CAD
CAAA14484-14 CAAA14484-22
Aluminum titanium oxide ≥99.5% (metals basis), powder -100 Mesh
Aluminum titanium oxide

Aluminum titanium oxide is used in refractory industry.


Purity: 99.5% CAS No.: 12004-39-6 Molecular Formula: Al2TiO5, may contain small amounts of other phases Formula Weight: 181.86 Physical Form: -100 Mesh Powder Odor: Odorless MDL No.: MFCD00064749

Formula: Al₂O₅Ti
Storage Temperature: Ambient
MDL Number: MFCD00064749
CAS Number: 12004-39-6
EINECS: 234-456-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR