Strontium isopropoxide ≥96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Strontium i-propoxide

14481-03 14481-06 14481-14
CAAA14481-03EA 76 CAD
CAAA14481-03 CAAA14481-06 CAAA14481-14
Strontium isopropoxide ≥96%
Strontium isopropoxide

Purity: 96% CAS No.: 88863-33-6 Molecular Formula: Sr[OCH(CH3)2]2 Formula Weight: 205.82 Physical Form: Powder Sensitivity: Air and Moisture Sensitive UN No.: 3205 MDL No.: MFCD00050487

Formula: C₆H₁₄O₂Sr
Melting Pt: 130 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00050487
CAS Number: 88863-33-6
UN: 3205
ADR: 4.2,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR