Tris(cyclopentadienyl)yttrium(III) ≥99.9%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tris(cyclopentadienyl)yttrium
14478-02
14478-04
CAAA14478-02EA
156.1
CAD
CAAA14478-02
CAAA14478-04
Tris(cyclopentadienyl)yttrium(III) ≥99.9%
Tris(cyclopentadienyl)yttrium(III)
Purity: 99.9%
CAS No.: 1294-07-1
Molecular Formula: C15H15Y
Formula Weight: 284.20
Physical Form: Powder, Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Melting Point: 295°
Boiling Point: 200° subl. (vacuum)
UN No.: 3181
MDL No.: MFCD00058706
Formula:
C₁₅H₁₅Y MW: 284.19 g/mol Boiling Pt: 200 °C (subl. in vacuum) Melting Pt: 295 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058706 CAS Number: 1294-07-1 EINECS: 215-069-4 UN: 3181 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...