Yttrium(III) isopropoxide ≥90%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14477-03
14477-06
CAAA14477-03EA
153.1
CAD
CAAA14477-03
CAAA14477-06
Yttrium(III) isopropoxide ≥90%
Yttrium(III) isopropoxide
Soluble in hexane, THF, toluene
Purity: 90+%
CAS No.: 2172-12-5
Molecular Formula: Y[OCH(CH3)2]3
Formula Weight: 266.17
Physical Form: Powder
Sensitivity: Moisture Sensitive
Melting Point: 160°
Boiling Point: 200-210°/1mm subl.
Solubility: Soluble in hexane, THF, toluene
UN No.: 2925
MDL No.: MFCD00015641
Formula:
C₉H₂₁O₃Y Boiling Pt: 200…210 °C (1 mmHg) (subl.) Melting Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015641 CAS Number: 2172-12-5 UN: 2925 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...